missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cis-1,2-cyclopropanedicarboxylate, 97+%, Thermo Scientific™

Catalog No. p-3736293
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC255340050 5g Glass bottle
AC255340250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC255340050 Supplier Thermo Scientific Chemicals Supplier No. 255340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 826-34-6
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.16
MDL Number MFCD00062805
InChI Key JBVOSZYUSFDYIN-SYDPRGILSA-N
Synonym dimethyl cis-1,2-cyclopropanedicarboxylate, dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, cis-dimethyl cyclopropane-1,2-dicarboxylate, dimethylcis-1,2-cyclopropanedicarboxylate, cyclopropane-1,2-dicarboxylic acid dimethyl ester, z, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s-rel, 1,2-dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, 1r,2s-dimethyl cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s, 1,2-cyclopropanedicarboxylicacid, 1,2-dimethyl ester, 1r,2s-rel
PubChem CID 642979
IUPAC Name dimethyl (1S,2R)-cyclopropane-1,2-dicarboxylate
SMILES COC(=O)C1CC1C(=O)OC

Specifications

Density 1.1500g/mL
Boiling Point 112.0°C (18.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4445 to 1.4465
Quantity 5g
Specific Gravity 1.15
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in all organic solvents
Formula Weight 158.16
Percent Purity 97+%
Chemical Name or Material Dimethyl cis-1,2-cyclopropanedicarboxylate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

RUO – Research Use Only