Learn More
Diethyl chloromalonate, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 114230250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Diethyl chloromalonate | |
1.2040g/mL | |
Authentic | |
C7H11ClO4 | |
1.431 | |
1.204 | |
WLWCQKMQYZFTDR-UHFFFAOYSA-N | |
diethyl 2-chloropropanedioate | |
84182 | |
90% |
14064-10-9 | |
113°C | |
88% min. (GC) | |
C2H5O2CCH(Cl)CO2C2H5 | |
25mL | |
diethyl chloromalonate, diethyl 2-chloromalonate, ethyl chloromalonate, ethylmonochloromalonate, propanedioic acid, chloro-, diethyl ester, chloromalonic acid, diethyl ester, diethyl chloropropanedioate, malonic acid, chloro-, diethyl ester, 1,3-diethyl 2-chloropropanedioate, propanedioic acid, 2-chloro-, 1,3-diethyl ester | |
CCOC(=O)C(C(=O)OCC)Cl | |
194.61 | |
194.61 |
Chemical Identifiers
14064-10-9 | |
194.61 | |
diethyl chloromalonate, diethyl 2-chloromalonate, ethyl chloromalonate, ethylmonochloromalonate, propanedioic acid, chloro-, diethyl ester, chloromalonic acid, diethyl ester, diethyl chloropropanedioate, malonic acid, chloro-, diethyl ester, 1,3-diethyl 2-chloropropanedioate, propanedioic acid, 2-chloro-, 1,3-diethyl ester | |
diethyl 2-chloropropanedioate |
C7H11ClO4 | |
WLWCQKMQYZFTDR-UHFFFAOYSA-N | |
84182 | |
CCOC(=O)C(C(=O)OCC)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 237-913-
RUO â Research Use Only