missing translation for 'onlineSavingsMsg'
Learn More

Diethyl chloromalonate, 90%, Thermo Scientific™

Chemical Identifiers

CAS 14064-10-9
Molecular Formula C7H11ClO4
Molecular Weight (g/mol) 194.61
InChI Key WLWCQKMQYZFTDR-UHFFFAOYSA-N
Synonym diethyl chloromalonate, diethyl 2-chloromalonate, ethyl chloromalonate, ethylmonochloromalonate, propanedioic acid, chloro-, diethyl ester, chloromalonic acid, diethyl ester, diethyl chloropropanedioate, malonic acid, chloro-, diethyl ester, 1,3-diethyl 2-chloropropanedioate, propanedioic acid, 2-chloro-, 1,3-diethyl ester
PubChem CID 84182
IUPAC Name diethyl 2-chloropropanedioate
SMILES CCOC(=O)C(C(=O)OCC)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC114230050
SDS View Documents
Thermo Scientific Chemicals
114230050
5mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC114230250
SDS View Documents
Thermo Scientific Chemicals
114230250
25mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

14064-10-9
194.61
diethyl chloromalonate, diethyl 2-chloromalonate, ethyl chloromalonate, ethylmonochloromalonate, propanedioic acid, chloro-, diethyl ester, chloromalonic acid, diethyl ester, diethyl chloropropanedioate, malonic acid, chloro-, diethyl ester, 1,3-diethyl 2-chloropropanedioate, propanedioic acid, 2-chloro-, 1,3-diethyl ester
diethyl 2-chloropropanedioate
C7H11ClO4
WLWCQKMQYZFTDR-UHFFFAOYSA-N
84182
CCOC(=O)C(C(=O)OCC)Cl

Specifications

14064-10-9
113°C
88% min. (GC)
C2H5O2CCH(Cl)CO2C2H5
5mL
diethyl chloromalonate, diethyl 2-chloromalonate, ethyl chloromalonate, ethylmonochloromalonate, propanedioic acid, chloro-, diethyl ester, chloromalonic acid, diethyl ester, diethyl chloropropanedioate, malonic acid, chloro-, diethyl ester, 1,3-diethyl 2-chloropropanedioate, propanedioic acid, 2-chloro-, 1,3-diethyl ester
CCOC(=O)C(C(=O)OCC)Cl
194.61
194.61
Diethyl chloromalonate
1.2040g/mL
Authentic
C7H11ClO4
1.431
1.204
WLWCQKMQYZFTDR-UHFFFAOYSA-N
diethyl 2-chloropropanedioate
84182
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 237-913-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only