missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 2,2-Diethoxyethylphosphonate, TCI Americaâ„¢
$177.63 - $512.13
Chemical Identifiers
CAS | 7598-61-0 |
---|---|
Molecular Formula | C10H23O5P |
Molecular Weight (g/mol) | 254.263 |
MDL Number | MFCD00009236 |
InChI Key | LUQYELQXRPNKRY-UHFFFAOYSA-N |
PubChem CID | 82071 |
IUPAC Name | 2-diethoxyphosphoryl-1,1-diethoxyethane |
SMILES | CCOC(CP(=O)(OCC)OCC)OCC |
Chemical Identifiers
7598-61-0 | |
254.263 | |
LUQYELQXRPNKRY-UHFFFAOYSA-N | |
2-diethoxyphosphoryl-1,1-diethoxyethane |
C10H23O5P | |
MFCD00009236 | |
82071 | |
CCOC(CP(=O)(OCC)OCC)OCC |
Specifications
7598-61-0 | |
130°C | |
MFCD00009236 | |
diethyl 2,2-diethoxyethylphosphonate, diethyl 2,2-diethoxyethyl phosphonate, diethyl2,2-diethoxyethylphosphonate, 2,2-diethoxyethylphosphonic acid diethyl ester, phosphonic acid, 2,2-diethoxyethyl-, diethyl ester, 2,2-diethoxyethyl diethoxyphosphino-1-one, diethyl phosphonacetaldehyde diethyl acetal, diethylphosphonoacetaldehyde diethylacetal, 2-diethoxyphosphoryl-1,1-diethoxy-ethane | |
CCOC(CP(=O)(OCC)OCC)OCC | |
254.263 | |
254.26 | |
Liquid |
Yellow | |
C10H23O5P | |
5 g | |
LUQYELQXRPNKRY-UHFFFAOYSA-N | |
2-diethoxyphosphoryl-1,1-diethoxyethane | |
82071 | |
≥96.0% (GC) | |
Diethyl 2,2-Diethoxyethylphosphonate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator