missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 2,2-Diethoxyethylphosphonate, TCI America™
Supplier: TCI America D242325G
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Diethyl 2,2-Diethoxyethylphosphonate | |
Yellow | |
C10H23O5P | |
25 g | |
LUQYELQXRPNKRY-UHFFFAOYSA-N | |
2-diethoxyphosphoryl-1,1-diethoxyethane | |
82071 | |
≥96.0% (GC) |
7598-61-0 | |
130°C | |
MFCD00009236 | |
diethyl 2,2-diethoxyethylphosphonate, diethyl 2,2-diethoxyethyl phosphonate, diethyl2,2-diethoxyethylphosphonate, 2,2-diethoxyethylphosphonic acid diethyl ester, phosphonic acid, 2,2-diethoxyethyl-, diethyl ester, 2,2-diethoxyethyl diethoxyphosphino-1-one, diethyl phosphonacetaldehyde diethyl acetal, diethylphosphonoacetaldehyde diethylacetal, 2-diethoxyphosphoryl-1,1-diethoxy-ethane | |
CCOC(CP(=O)(OCC)OCC)OCC | |
254.263 | |
254.26 | |
Liquid |
Chemical Identifiers
7598-61-0 | |
254.263 | |
LUQYELQXRPNKRY-UHFFFAOYSA-N | |
2-diethoxyphosphoryl-1,1-diethoxyethane |
C10H23O5P | |
MFCD00009236 | |
82071 | |
CCOC(CP(=O)(OCC)OCC)OCC |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator