missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 1,1-Cyclohexanedicarboxylate 95.0+%, TCI America™
Supplier: TCI America C164025G
Specifications
Diethyl 1,1-Cyclohexanedicarboxylate | |
Colorless | |
C12H20O4 | |
25 g | |
MLHUKQNAQSRVKI-UHFFFAOYSA-N | |
1,1-diethyl cyclohexane-1,1-dicarboxylate | |
249270 | |
≥95.0% (GC) |
1139-13-5 | |
137°C | |
MFCD02093457 | |
diethyl 1,1-cyclohexanedicarboxylate, 1,1-cyclohexanedicarboxylic acid diethyl ester, 1,1-diethyl cyclohexane-1,1-dicarboxylate, cyclohexane-1,1-dicarboxylic acid diethyl ester, 1,1-cyclohexanedicarboxylic acid, diethyl ester, acmc-1c6ic, 1,1-bis ethoxycarbonyl cyclohexane, cyclohexanedicarboxylicaciddiethylester, diethyl 1,1-cyclohexanedicarboxylate #, 1,1-cyclohexanedicarboxylicacid, 1,1-diethyl ester | |
CCOC(=O)C1(CCCCC1)C(=O)OCC | |
228.29 | |
228.29 | |
Liquid |
Chemical Identifiers
1139-13-5 | |
228.29 | |
MLHUKQNAQSRVKI-UHFFFAOYSA-N | |
249270 | |
CCOC(=O)C1(CCCCC1)C(=O)OCC |
C12H20O4 | |
MFCD02093457 | |
diethyl 1,1-cyclohexanedicarboxylate, 1,1-cyclohexanedicarboxylic acid diethyl ester, 1,1-diethyl cyclohexane-1,1-dicarboxylate, cyclohexane-1,1-dicarboxylic acid diethyl ester, 1,1-cyclohexanedicarboxylic acid, diethyl ester, acmc-1c6ic, 1,1-bis ethoxycarbonyl cyclohexane, cyclohexanedicarboxylicaciddiethylester, diethyl 1,1-cyclohexanedicarboxylate #, 1,1-cyclohexanedicarboxylicacid, 1,1-diethyl esterShow More | |
1,1-diethyl cyclohexane-1,1-dicarboxylate |
Safety and Handling
TSCA : No