missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,1-Cyclohexanedicarboxylate 95.0+%, TCI America™

Catalog No. p-7125098
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16401G 1 g
C16405G 5 g
C164025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C16401G Supplier TCI America Supplier No. C16401G
Only null left

Chemical Identifiers

CAS 1139-13-5
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.29
MDL Number MFCD02093457
InChI Key MLHUKQNAQSRVKI-UHFFFAOYSA-N
Synonym diethyl 1,1-cyclohexanedicarboxylate, 1,1-cyclohexanedicarboxylic acid diethyl ester, 1,1-diethyl cyclohexane-1,1-dicarboxylate, cyclohexane-1,1-dicarboxylic acid diethyl ester, 1,1-cyclohexanedicarboxylic acid, diethyl ester, acmc-1c6ic, 1,1-bis ethoxycarbonyl cyclohexane, cyclohexanedicarboxylicaciddiethylester, diethyl 1,1-cyclohexanedicarboxylate #, 1,1-cyclohexanedicarboxylicacid, 1,1-diethyl ester
PubChem CID 249270
IUPAC Name 1,1-diethyl cyclohexane-1,1-dicarboxylate
SMILES CCOC(=O)C1(CCCCC1)C(=O)OCC

Specifications

Color Colorless
Boiling Point 137°C
Quantity 1 g
Formula Weight 228.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 1,1-Cyclohexanedicarboxylate