missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Myo-inositol, 99.26%, For HPLC analysis, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210205290
Specifications
D-Myo-inositol | |
87-89-8 | |
6.2g/mL | |
C6H12O6 | |
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol | |
C1(C(C(C(C(C1O)O)O)O)O)O | |
180.156 | |
CHEBI:24848 | |
Powder |
For HPLC analysis | |
White | |
99.26% | |
500 g | |
CDAISMWEOUEBRE-UHFFFAOYSA-N | |
cyclohexane-1,2,3,4,5,6-hexol | |
892 | |
180.2 |
Chemical Identifiers
87-89-8 | |
180.156 | |
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol | |
CHEBI:24848 | |
C1(C(C(C(C(C1O)O)O)O)O)O |
C6H12O6 | |
CDAISMWEOUEBRE-UHFFFAOYSA-N | |
892 | |
cyclohexane-1,2,3,4,5,6-hexol |