missing translation for 'onlineSavingsMsg'
Learn More

D-Myo-inositol, 99.26%, For HPLC analysis, MP Biomedicals™

Catalog No. p-4405728
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10205250 50 g
ICN10205280 100 g
ICN10205290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10205250 Supplier MP Biomedicals Inc Supplier No. 0210205250
Only null left

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Color White
Density 6.2g/mL
Assay Percent Range 99.26%
Quantity 50 g
Formula Weight 180.2
Physical Form Powder
Chemical Name or Material D-Myo-inositol, For HPLC analysis