missing translation for 'onlineSavingsMsg'
Learn More

CPC™ Plastic Quick-Disconnect Fittings DFS Item

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables tests.

Supplier:  CPC™ MPC17004T03

 View more versions of this product

Catalog No. 05-113-143

Only null left
Explore available promotions

Description

Description

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables

CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.

  • Material: Medical-grade ABS, polycarbonate and polysulfone
  • Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
  • Fow Rate: Cv∽5.5 (79.25) max.

  • Ergonomic thumb latch
  • Audible 'click' when connected
  • USP Class VI materials
  • Can be sterilized by autoclave, EtO, e-beam, or gamma
  • Coupling halves can be rotated when connected
  • Tapered barb
  • BSE/TSE free materials

  • Cell culture media packaging and transfer lines
  • Harvest and collection applications
  • Buffer solutions
  • Tube sets
  • Urology drainage/collection systems
  • Hypo/hyperthermia blankets

Specifications

Specifications

USP Class VI, ADCF
0.25 in.
Polycarbonate
28 mm
Cell culture media packaging and transfer lines, harvest and collection applications
-40°C
0.96 in.
250°F
Hose Barb Non-valved In-line Coupling Body
Autoclavable
6.4 mm
1.1 in.
6.4mm
-40°F
25 mm
Purple
121°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions