missing translation for 'onlineSavingsMsg'
Learn More

CPC™ Plastic Quick-Disconnect Fittings

Numéro de catalogue. p-3850952
Encompass
Change view
Click to view available options
Diameter (Metric) Inner:
6.4 mm
9.5 mm
Product Type:
Hose Barb Non-valved In-line Coupling Body
Material:
Polycarbonate
Polysulfone
Color:
Amber
Purple
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Diameter (Metric) Inner Product Type Material Color
05113143 6.4 mm Hose Barb Non-valved In-line Coupling Body Polycarbonate Purple
05113145 6.4 mm Hose Barb Non-valved In-line Coupling Body Polysulfone Amber
05113101 9.5 mm Hose Barb Non-valved In-line Coupling Body Polysulfone Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 05113143 Fournisseur CPC™ Code fournisseur MPC17004T03
Il en reste null

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables tests.

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables

CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.

  • Material: Medical-grade ABS, polycarbonate and polysulfone
  • Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
  • Fow Rate: Cv∽5.5 (79.25) max.

  • Ergonomic thumb latch
  • Audible 'click' when connected
  • USP Class VI materials
  • Can be sterilized by autoclave, EtO, e-beam, or gamma
  • Coupling halves can be rotated when connected
  • Tapered barb
  • BSE/TSE free materials

  • Cell culture media packaging and transfer lines
  • Harvest and collection applications
  • Buffer solutions
  • Tube sets
  • Urology drainage/collection systems
  • Hypo/hyperthermia blankets

Spécifications

Certifications/Compliance USP Class VI, ADCF
Autoclavable Autoclavable
Diameter (English) Inner 0.25 in.
Diameter (Metric) Inner 6.4 mm
Material Polycarbonate
Overall Length (English) 1.1 in.
Overall Length (Metric) 28 mm
For Use With 6.4mm
For Use With (Application) Cell culture media packaging and transfer lines, harvest and collection applications
Min. Operating Temperature (English) -40°F
Min. Operating Temperature (Metric) -40°C
Height (Metric) 25 mm
Height (English) 0.96 in.
Color Purple
Max. Operating Temperature (English) 250°F
Max. Operating Temperature (Metric) 121°C
Product Type Hose Barb Non-valved In-line Coupling Body
Afficher plus de résultats Afficher moins de résultats