Corning™ Costar™ Netwell™ Reagent Trays, Non-Sterile
Specially designed Netwell™ carriers with handles allow simultaneous processing of up to 12 samples per carrier
Supplier: Corning™ 3519
Description
- Specially designed Netwell carriers and handles allow simultaneous processing of up to 12 samples per carrier
- Polystyrene reagent trays are available in white for colorimetric reaction contrast, or black for better visibility of tissue sections
- Each carrier kit contains eight carriers and eight handles
Specifications
Tray | |
25/Pk. | |
200/Cs. | |
White |