Learn More
Corning™ Costar™ Netwell™ Carrier Kit, Non-Sterile
Allows simultaneous processing
Supplier:  Corning™ C3521
 View more versions of this product
Catalog No. 07-200-224
Only null left
Explore available promotions
Description
Description
  • These specially designed Netwell carriers and handles allow simultaneous processing of up to 12 Netwell inserts per carrier
  • Each carrier kit contains eight carriers and eight handles
  • Greener Choice Claims
    • Environmental benefits include:
      • Source Reduction
    Specifications
    Specifications
    Carrier Kit
    24 mm
    8/Cs.
    Allows simultaneous processing of up to 6 samples per carrier
    8/Pk.
    Product Suggestions
    Product Suggestions