missing translation for 'onlineSavingsMsg'
Learn More

Corningā„¢ Universal CellSTACKā„¢ Caps

For filling, venting, and manipulation

Supplier:  Corningā„¢ 3339

 View more versions of this product

Catalog No. 07-202-522


Only null left
Explore available promotions

Description

Description

  • Choice of 33mm polyethylene, solid or vented
  • Sterilized by gamma irradiation

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

Orange
3.3 cm
Male MPC Polycarbonate with a 0.95cm I.D. coupling and a female MPC Polycarbonate end cap
Single-use
Screw
HDPE
For filling, venting, and manipulation
Cap, Filling
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions