Learn More
Corning™ Filtered IsoTip™ Universal Fit Racked Pipet Tips
GREENER_CHOICE
Ideal for applications where avoiding cross-contamination is critical, such as DNA amplification and radioisotope handling
Supplier:  Corning™ 4807
 View more versions of this product
Catalog No. 07-200-501
Only null left
Explore available promotions
Description
Description
  • IsoTip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolized contaminants from coming in contact with pipettor shafts. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website.)
  • Ideal for applications where avoiding cross contamination is critical, such as DNA amplification and radioisotope handling.
  • Packaged sterile.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
0.2 to 10 ÎĽL
Sterile
Rack
Rack
Isotip
Filtered
Natural
Round
Gilson™ Pipetman™, Universal Fit Ultramicro
Polypropylene
Product Suggestions
Product Suggestions