missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulfate Solution for Aluminum Can Coating Test, Reagents

Copper Sulfate Solution for Aluminum Can Coating Test, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$304.33 - $329.10
Specifications
Boiling Point | 102°C |
---|---|
Chemical Name or Material | Copper Sulfate Solution for Aluminum Can Coating Test |
CAS | 7732-18-5,7758-99-8 |
Color | Blue/Green |
pH | 1.98 to 2.16 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
01-335-668
|
Labchem Inc
CC1021004A |
4 L | Poly Bottle |
Each for $304.33
|
|
|||||
01-335-667
|
Labchem Inc
CC1021004C |
4 L | Glass Bottle |
Each for $329.10
|
|
|||||
Specifications
102°C | |
7732-18-5,7758-99-8 | |
1.98 to 2.16 | |
4 L | |
Copper(II) Sulfate | |
1.1 |
Copper Sulfate Solution for Aluminum Can Coating Test | |
Blue/Green | |
Liquid | |
CuSO4/H2O | |
Reagent |