missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate Solution for Aluminum Can Coating Test, Reagents

Catalog No. 01335668
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01335668 4 L Poly Bottle
01335667 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335668 Supplier Labchem Inc Supplier No. CC1021004A
Only null left

Copper Sulfate Solution for Aluminum Can Coating Test, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Copper Sulfate Solution for Aluminum Can Coating Test
Boiling Point 102°C
Color Blue/Green
CAS 7732-18-5, 7758-99-8
pH 1.98 to 2.16
Physical Form Liquid
Quantity 4 L
Molecular Formula CuSO4/H2O
Synonym Copper(II) Sulfate
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1
Show More Show Less