missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Color Standard, Pt-Co, 500 Units APHA, Stock Solution, Certified, LabChem™
Supplier: LabChem LC133002
Specifications
Color Standard | |
Amber Glass | |
16921-30-5,7791-13-1,7647-01-0,7732-18-5 | |
Liquid | |
UN1789 | |
Soluble in water | |
485.99 |
Pt-Co, 500 Units APHA, Stock Solution | |
95.43,4.34,0.13,0.1 | |
Passes Test | |
K2(PtCl6) | |
Orange | |
1 L | |
Certified |