missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citrate-Cyanide Reducing Solution, For Lead, LabChem™
Supplier: LabChem LC131301
Specifications
Citrate-Cyanide Reducing Solution | |
For Lead | |
66.7,17.64,13.33,1.33,0.67 | |
UN1935 | |
65.12 | |
Poly Bottle | |
500 mL |
Colorless to Yellow | |
1336-21-6,7732-18-5,3012-65-5,151-50-8,7757-83-7 | |
KCN | |
Soluble in water | |
Passes Test | |
Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Gaseous Ammonia | |
Liquid |