missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexene, 97%

CAS: 7642-09-3 | C6H12 | 84.16 g/mol

Supplier:  Thermo Scientific Chemicals H5348514

 View more versions of this product

Catalog No. AAH5348514


Only null left
Explore available promotions

Description

Description

cis-3-Hexene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-3-Hexene is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

cis-3-Hexene
0.681
-25°C (-13°F)
1.395
25 g
1718858
97%
7642-09-3
66°C to 68°C
C6H12
MFCD00085300
UN3295
84.16

Chemical Identifiers

7642-09-3
MFCD00085300
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H225-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only