accessibility menu, dialog, popup

UserName

CH3CH3(Z)

CAS RN 7642-09-3

IUPAC Name: (3Z)-hex-3-ene
Synonyms: (3Z)-hex-3-ene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: ZQDPJFUHLCOCRG-WAYWQWQTSA-N
SMILES: CC\C=C/CC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)