Learn More
cis-3-Hexen-1-ol, 98%
CAS: 928-96-1 | C6H12O | 100.16 g/mol
Supplier: Thermo Scientific Chemicals 120770100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
cis-3-Hexen-1-ol | |
Colorless to Yellow | |
156.0°C to 157.0°C | |
Authentic | |
Glass bottle | |
1.4380 to 1.4410 | |
MFCD00063217 | |
01, I, 229 | |
10, 4592 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and propylene glycol, miscible with oils | |
CC\C=C/CCO | |
100.16 | |
CHEBI:28857 | |
98% |
928-96-1 | |
0.8480g/mL | |
59°C | |
97.5% min. (GC) | |
C6H12O | |
C2H5CH=CHCH2CH2OH | |
10 mL | |
0.848 | |
z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z | |
UFLHIIWVXFIJGU-ARJAWSKDSA-N | |
(3Z)-hex-3-en-1-ol | |
5281167 | |
100.16 | |
Liquid |
Chemical Identifiers
928-96-1 | |
100.16 | |
UFLHIIWVXFIJGU-ARJAWSKDSA-N | |
5281167 | |
CC\C=C/CCO |
C6H12O | |
MFCD00063217 | |
z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, zShow More | |
CHEBI:28857 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advic
GHS Signal Word: Warning
EINECSNumber : 213-192-8
RUO – Research Use Only