missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexen-1-ol, 98%

CAS: 928-96-1 | C6H12O | 100.16 g/mol

$91.83 - $271.61

Chemical Identifiers

CAS 928-96-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00063217
InChI Key UFLHIIWVXFIJGU-ARJAWSKDSA-N
Synonym z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z
PubChem CID 5281167
ChEBI CHEBI:28857
SMILES CC\C=C/CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC120770100
View Documents Promotion Details
N/A 10 mL Glass bottle
Each for $91.83
Only null left
 
AC120770500
View Documents Promotion Details
N/A 50 mL Glass bottle
Each for $271.61
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

928-96-1
100.16
UFLHIIWVXFIJGU-ARJAWSKDSA-N
5281167
CC\C=C/CCO
C6H12O
MFCD00063217
z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z
CHEBI:28857

Specifications

928-96-1
0.8480g/mL
59°C
97.5% min. (GC)
1.4380 to 1.4410
MFCD00063217
0.848
z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z
UFLHIIWVXFIJGU-ARJAWSKDSA-N
(3Z)-hex-3-en-1-ol
5281167
100.16
Liquid
Colorless to Yellow
156.0°C to 157.0°C
Authentic
C6H12O
C2H5CH=CHCH2CH2OH
01, I, 229
10, 4592
Solubility in water: insoluble. Other solubilities: soluble in alcohol and propylene glycol, miscible with oils
CC\C=C/CCO
100.16
CHEBI:28857
98%
cis-3-Hexen-1-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advic

GHS Signal Word: Warning

EINECSNumber : 213-192-8

SDS
Documents

Documents

RUO – Research Use Only