Learn More
cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%
CAS: 24947-68-0 | C7H16NO | 130.21 g/mol
$421.68 - $789.70
Chemical Identifiers
CAS | 24947-68-0 |
---|---|
Molecular Formula | C7H16NO |
Molecular Weight (g/mol) | 130.21 |
MDL Number | MFCD00143984 |
InChI Key | JGKFBZBVCAWDFD-BQBZGAKWSA-O |
Synonym | 1s,2s-2-hydroxycyclohexyl methanaminium |
PubChem CID | 2724656 |
IUPAC Name | (1S,2S)-2-(aminomethyl)cyclohexan-1-ol;hydrochloride |
SMILES | [NH3+]C[C@@H]1CCCC[C@@H]1O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC265882500
|
Thermo Scientific Chemicals
265882500 |
250 mg | Glass bottle |
Each for $421.68
|
|
|||||
AC265880010
|
Thermo Scientific Chemicals
265880010 |
1 g | Glass bottle |
Each for $789.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24947-68-0 | |
130.21 | |
JGKFBZBVCAWDFD-BQBZGAKWSA-O | |
2724656 | |
[NH3+]C[C@@H]1CCCC[C@@H]1O |
C7H16NO | |
MFCD00143984 | |
1s,2s-2-hydroxycyclohexyl methanaminium | |
(1S,2S)-2-(aminomethyl)cyclohexan-1-ol;hydrochloride |
Specifications
24947-68-0 | |
White | |
98.5% min. (Argentometry) | |
C7H16NO | |
250 mg | |
Solubility in water: soluble. Other solubilities: soluble in methanol | |
[NH3+]C[C@@H]1CCCC[C@@H]1O | |
130.21 | |
165.66 | |
Crystalline Powder |
182.0°C to 185.0°C | |
Authentic | |
Glass bottle | |
MFCD00143984 | |
1s,2s-2-hydroxycyclohexyl methanaminium | |
JGKFBZBVCAWDFD-BQBZGAKWSA-O | |
(1S,2S)-2-(aminomethyl)cyclohexan-1-ol;hydrochloride | |
2724656 | |
99% | |
cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only