missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%

CAS: 24947-68-0 | C7H16NO | 130.21 g/mol

Supplier:  Thermo Scientific Chemicals 265880010

 View more versions of this product

Catalog No. AC265880010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

cis-2-Aminomethyl-1-cyclohexanol hydrochloride
24947-68-0
White
98.5% min. (Argentometry)
C7H16NO
1 g
Solubility in water: soluble. Other solubilities: soluble in methanol
[NH3+]C[C@@H]1CCCC[C@@H]1O
130.21
165.66
Crystalline Powder
99%
182.0°C to 185.0°C
Authentic
Glass bottle
MFCD00143984
1s,2s-2-hydroxycyclohexyl methanaminium
JGKFBZBVCAWDFD-BQBZGAKWSA-O
(1S,2S)-2-(aminomethyl)cyclohexan-1-ol;hydrochloride
2724656
99%

Chemical Identifiers

24947-68-0
130.21
JGKFBZBVCAWDFD-BQBZGAKWSA-O
2724656
[NH3+]C[C@@H]1CCCC[C@@H]1O
C7H16NO
MFCD00143984
1s,2s-2-hydroxycyclohexyl methanaminium
(1S,2S)-2-(aminomethyl)cyclohexan-1-ol;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only