missing translation for 'onlineSavingsMsg'
Learn More

Cesium nitrate, 99.8% (metals basis)

Preparation of Cs compounds | CAS: 7789-18-6 | CsNO3 | 194.909 g/mol

Supplier:  Thermo Scientific Chemicals 01288418

 View more versions of this product

Catalog No. AA1288418


Only null left
Explore available promotions

Description

Description

Preparation of cesium compoundsCesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P220-P221-P264-P270-P272-P280-P301+P312-P308+P313-P330-P370+P378q-P370+P380+P375-P372-P501c

H271-H302-H361f

DOTInformation : Hazard Class: 5.1; Packaging Group: III

EINECSNumber : 232-146-8

RTECSNumber : FL0700000

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only