missing translation for 'onlineSavingsMsg'
Learn More

Cesium nitrate, 99.8% (metals basis)

Catalog No. p-3256980
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1288418 50 g
AA1288430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1288418 Supplier Thermo Scientific Chemicals Supplier No. 01288418
Only null left

Preparation of Cs compounds | CAS: 7789-18-6 | CsNO3 | 194.909 g/mol

Preparation of cesium compoundsCesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.909
MDL Number MFCD00010963
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
Synonym cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]

Specifications

Boiling Point (decomposition)
Melting Point 414°C
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99.8% (metals basis)
Solubility Information Soluble in 5 parts cold and 0.5 parts boiling water. Soluble in acetone. Slightly soluble in alcoholSoluble in water and acetone. Slightly soluble in ethanol.
Formula Weight 194.91
Refractive Index 1.55
Sensitivity Hygroscopic
Density 3.68
Chemical Name or Material Cesium nitrate
Show More Show Less
Hazard Category H271-H302-H361f
Precautionary Statement P201-P202-P220-P221-P264-P270-P272-P280-P301+P312-P308+P313-P330-P370+P378q-P370+P380+P375-P372-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 232-146-8
RTECSNumber FL0700000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only