missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride, 99.999%, (Solutions are clear and colorless/Ultra Pure ), MP Biomedicals
Especially suited for critical density gradient techniques
Supplier: MP Biomedicals Inc 0215058980
Specifications
Cesium Chloride | |
7647-17-8 | |
100 g | |
ClCs | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
[Cl-].[Cs+] | |
168.36 | |
CHEBI:63039 | |
Ultrapure |
Colorless | |
Solution | |
99.999% | |
MFCD00010955 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
caesium(1+) chloride | |
24293 | |
168.4 |
Chemical Identifiers
7647-17-8 | |
168.36 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
24293 | |
[Cl-].[Cs+] |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chlorideShow More | |
CHEBI:63039 |
Safety and Handling
Recommended Storage : Room Temperature