missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium Chloride, 99.999%, (Solutions are clear and colorless/Ultra Pure ), MP Biomedicals

Especially suited for critical density gradient techniques

$691.58 - $691.58

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.355
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15058980
View Documents Promotion Details
Mp Biomedicals
0215058980
100g
Each for $691.58
 
Description

Description

Specifications

Chemical Identifiers

7647-17-8
168.355
caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
CHEBI:63039
[Cl-].[Cs+]
ClCs
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride

Specifications

7647-17-8
100g
ClCs
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.4
Ultrapure
Colorless
99.999%
caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
[Cl-].[Cs+]
168.355
CHEBI:63039
Solution
Cesium Chloride
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Product Certifications