Learn More
CERILLIANT™ Barbital Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore B0701ML
Catalog No. 01001056
Only null left
Explore available promotions
Description
Description
Barbital, also known as barbitone and marketed under the brand names Veronal and Medinal, was the first barbiturate commercially available. A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls in barbiturate LC/MS or GC/MS testing methods for clinical toxicology, urine drug testing or forensic analysis applications.
Specifications
Specifications
67-56-1,57-44-3
MFCD00036212
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C8H12N2O3
UN 1230 - class 3 - PG 2 - Methanol, solution
184.19
1 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation