missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD169 Monoclonal Antibody (ED3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Fisher Scientific MA180164
Description
For FACS analysis, use 10 ÎĽL of the suggested working dilution to label 1x10^6 cells in 100 ÎĽL. Mouse anti rat CD169 antibody, clone ED3 recognizes the rat CD169 cell surface antigen, a approximately 185 kDa molecule expressed by macrophages, predominately confined to lymphoid organs only.
The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.Specifications
CD169 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
SIGLEC1 | |
Rat Spleen cell homogenate. | |
250 ÎĽg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
ED3 | |
Unconjugated | |
SIGLEC1 | |
Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN | |
Mouse | |
Protein A | |
RUO | |
311426 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |