Learn More
MilliporeSigma™ Calbiochem™ Albumin, Human Serum, Fraction V, Low Heavy Metals
Prepared under non-denaturing conditions by modification of Cohn procedure
Supplier:  MilliporeSigma™ 126661GM
 View more versions of this product
Catalog No. 126661GM
Only null left
Explore available promotions
Description
Description
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Protein Standard, Protein Base or Filler, Stabilizer and Diluent in Serology and Immunohematology
Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Solid
1 g
2° to 8°C
≥95% by SDS PAGE
Product Suggestions
Product Suggestions