missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl N-(3-Trifluoromethylphenyl)anthranilate 98.0+%, TCI Americaâ„¢
$141.73 - $421.41
Chemical Identifiers
CAS | 67330-25-0 |
---|---|
Molecular Formula | C18H18F3NO2 |
Molecular Weight (g/mol) | 337.342 |
MDL Number | MFCD00866001 |
InChI Key | JDLSRXWHEBFHNC-UHFFFAOYSA-N |
Synonym | N-(3-Trifluoromethylphenyl)anthranilic Acid Butyl Ester, Butyl Flufenamate, Flufenamic Acid Butyl Ester, Butyl 2-[[3-(Trifluoromethyl)phenyl]amino]benzoate, 2-[[3-(Trifluoromethyl)phenyl]amino]benzoic Acid Butyl Ester, UfenamateShow More |
PubChem CID | 5632 |
IUPAC Name | butyl 2-[3-(trifluoromethyl)anilino]benzoate |
SMILES | CCCCOC(=O)C1=CC=CC=C1NC2=CC=CC(=C2)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
B5391200MG
|
TCI America
B5391200MG |
200 mg |
Each for $141.73
|
|
||||||
B53911G
|
TCI America
B53911G |
1 g |
Each for $421.41
|
|
||||||
Chemical Identifiers
67330-25-0 | |
337.342 | |
JDLSRXWHEBFHNC-UHFFFAOYSA-N | |
5632 | |
CCCCOC(=O)C1=CC=CC=C1NC2=CC=CC(=C2)C(F)(F)F |
C18H18F3NO2 | |
MFCD00866001 | |
N-(3-Trifluoromethylphenyl)anthranilic Acid Butyl Ester, Butyl Flufenamate, Flufenamic Acid Butyl Ester, Butyl 2-[[3-(Trifluoromethyl)phenyl]amino]benzoate, 2-[[3-(Trifluoromethyl)phenyl]amino]benzoic Acid Butyl Ester, UfenamateShow More | |
butyl 2-[3-(trifluoromethyl)anilino]benzoate |
Specifications
67330-25-0 | |
170°C | |
MFCD00866001 | |
N-(3-Trifluoromethylphenyl)anthranilic Acid Butyl Ester, Butyl Flufenamate, Flufenamic Acid Butyl Ester, Butyl 2-[[3-(Trifluoromethyl)phenyl]amino]benzoate, 2-[[3-(Trifluoromethyl)phenyl]amino]benzoic Acid Butyl Ester, Ufenamate | |
CCCCOC(=O)C1=CC=CC=C1NC2=CC=CC(=C2)C(F)(F)F | |
337.342 | |
337.34 | |
Liquid |
Yellow | |
C18H18F3NO2 | |
200 mg | |
JDLSRXWHEBFHNC-UHFFFAOYSA-N | |
butyl 2-[3-(trifluoromethyl)anilino]benzoate | |
5632 | |
≥98.0% (HPLC,N) | |
Butyl N-(3-Trifluoromethylphenyl)anthranilate |
Safety and Handling
RTECSNumber : DH8511000
TSCA : No