missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 7, Phosphate buffer
CAS: 7558-79-4 | H2KO4P
$71.50 - $298.40
Specifications
Chemical Name or Material | Buffer solution pH 7 |
---|---|
CAS | 7558-79-4,7732-18-5 |
Molecular Formula | H2KO4P |
MDL Number | MFCD03097944 |
Packaging | Plastic bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC258591000
|
Thermo Scientific Chemicals
258591000 |
100 mL |
Each for $71.50
|
|
||||||
AC258595000
|
Thermo Scientific Chemicals
258595000 |
500 mL |
Each for $99.70
|
|
||||||
AC258590010
|
Thermo Scientific Chemicals
258590010 |
1 L |
Each for $157.67
|
|
||||||
AC258590025
|
Thermo Scientific Chemicals
258590025 |
2.5 L |
Each for $298.40
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH bufferSpecifications
Buffer solution pH 7 | |
H2KO4P | |
Plastic bottle | |
14mmHg at 20°C |
7558-79-4,7732-18-5 | |
MFCD03097944 | |
14, 7659 | |
136.09 |
RUO – Research Use Only