missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4, Citrate buffer

CAS: 68-04-2 | C8H5KO4

Supplier:  Thermo Scientific Chemicals 258585000

 View more versions of this product

Catalog No. AC258585000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
Specifications

Specifications

Buffer solution pH 4
68-04-2,7647-14-5,7732-18-5
3.98 to 4.02 (20°C)
500 mL
MFCD00130806
Traceable to NIST
204.23
Phthalate buffer
Colorless
Liquid
C8H5KO4
Plastic bottle
Solubility in water: soluble.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only