missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ S 384-well Microplates
Non-treated sterile microplates for diverse applications
$972.81 - $3575.30
Specifications
Format | Standard |
---|---|
Lid | With Lid |
Array | 16 x 24 |
Material | Polystyrene |
No. of Wells | 384 |
Catalog Number | Mfr. No. | Color | Volume (Metric) Well | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Volume (Metric) Well | Price | Quantity | |||||
14-386-023
|
N/A | Black | 0.1 mL |
Pack of 50 Each for $1,178.63
|
|
|||||
14-386-060
|
N/A | Clear | 0.1 mL |
Pack of 50 Each for $972.81
|
|
|||||
14-386-022
|
N/A | White | 0.1 mL |
Pack of 50 Each for $1,115.07
|
|
|||||
14-386-028
|
N/A | White, Clear Bottom | 0.12 mL |
Pack of 50 Each for $3,575.30
|
|
|||||
14-386-029
|
N/A | Black, Clear Bottom | 0.12 mL |
Pack of 50 Each for $3,575.30
|
|
|||||
Description
- Non-treated PS surface
- Medium binding
- Sterilized via beta-radiation
- Sterile plate for most applications
Specifications
Standard | |
16 x 24 | |
384 | |
Non-treated | |
Laboratory tasks, such as homogeneous assays, screening and storage |
With Lid | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
50/Cs. |