Learn More
BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ S 384-well Microplates
Non-treated sterile microplates for diverse applications
 View more versions of this product
Catalog No. 14-386-060
Only null left
Explore available promotions
Description
Description
  • Non-treated PS surface
  • Medium binding
  • Sterilized via beta-radiation
  • Sterile plate for most applications
Specifications
Specifications
No Barcode
With Lid
Clear
0.1 mL
Sterile
Flat Bottom (F-well)
50/Cs.
Standard
16 x 24
Polystyrene
384
Non-treated
Laboratory tasks, such as homogeneous assays, screening and storage
Product Suggestions
Product Suggestions