Learn More
Brady™ Electrical Hazard Signs: DANGER - LIVE WIRE OVERHEAD
Identify electrical hazard and high voltage areas
Supplier:  Brady™ 84917
 View more versions of this product
Catalog No. 19-035-915
Only null left
Explore available promotions
Description
Description
  • Available in two materials - plastic and polyester
  • Available in different widths and heights
  • Black/red on white
Specifications
Specifications
ANSI Z535, OSHA 1910.145, PSTC-33,
1 Ea.
Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label
17.78 cm
-40°F
10 in.
-40°C to 110°C
DANGER Live Wire Overhead
Red on White
Polyester
Surface Mount
7 in.
230°F
0.006 to 0.01 in.
25.40 cm
-40°F to 230°F
Electrical Hazard Sign
Product Suggestions
Product Suggestions