missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Electrical Hazard Signs: DANGER - LIVE WIRE OVERHEAD
Identify electrical hazard and high voltage areas
$22.90
Specifications
| Quantity | 1 Ea. |
|---|---|
| Mounting Type | Surface Mount |
| Min. Temperature (English) | -40°F |
| Displayed Text | DANGER Live Wire Overhead |
| Product Type | Electrical Hazard Sign |
| Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity & Availability | |||
|
19035914
|
Brady™
BRD25557 |
Plastic | 10 in. | 14 in. |
Each for $22.90
|
|
|||
Description
- Available in two materials - plastic and polyester
- Available in different widths and heights
- Black/red on white
Specifications
| 1 Ea. | |
| -40°F | |
| Electrical Hazard Sign |
| Surface Mount | |
| DANGER Live Wire Overhead |