Learn More
Brady™ Danger: No Smoking, No Open Flames, No Sparks Signs
Danger: No Smoking, No Open Flames, No Sparks safety signs from Brady™ help to manage and sustain a clean and safe workplace and are available in a variety of materials to best fit your application.
Supplier:  Brady™ 88379
 View more versions of this product
Catalog No. 19-036192
Only null left
Explore available promotions
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Specifications
OSHA
1 Ea.
Self-sticking Polyester
10 in.
230°F
0.006 to 0.01 in.
35.5 cm
-40°F to 230°F
DANGER No Smoking No Open Flames No Sparks
Red on White
Polyester
Permanent Acrylic
Safety sign, identification of fire alarm, extinguisher and emergency exit
25.4 cm
-40°F
14 in.
-40°C to 110°C
10 x 14 in.
No Smoking Sign
Product Suggestions
Product Suggestions