Learn More
Brady™ BradyGlo™ Glow-in-the-Dark Exit and Directional Sign: NO EXIT
Door, passage/stairway w/no exit/way to get to exit, but could be exit must be marked with this sign
Supplier:  Brady™ 85481
 View more versions of this product
Catalog No. 19-000-451
Only null left
Explore available promotions
Description
Description
  • Photoluminescent sign
  • Ensures safe well-lit exit during evacuation
  • BradyGlo™ self sticking polyester
  • Red on green
  • English
  • 17.78 x 25.40cm (7 x 10 in.)
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Plastic
25.4 cm
35.5 cm
10 x 14 in.
Exit and Directional Sign (Glow-in-the-Dark)
10 in.
14 in.
English
NO EXIT
Red on Glow
Product Suggestions
Product Suggestions