missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOC-N-Methyl-L-alanine, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 442930050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
BOC-N-Methyl-L-alanine | |
99% | |
5g | |
boc-n-me-ala-oh, boc-n-methyl-l-alanine, n-boc-n-methyl-l-alanine, s-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-l-alanine, boc-n-a-methyl-l-alanine, s-2-tert-butoxycarbonyl-methyl-amino-propionic acid, l-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino propanoic acid, 2s-2-tert-butoxy carbonyl methyl amino propanoic acid | |
CC(C(=O)O)N(C)C(=O)OC(C)(C)C | |
203.24 | |
203.24 |
16948-16-6 | |
C9H17NO4 | |
− 29.80 | |
VLHQXRIIQSTJCQ-LURJTMIESA-N | |
(2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid | |
7009590 | |
99% |
Chemical Identifiers
16948-16-6 | |
203.24 | |
boc-n-me-ala-oh, boc-n-methyl-l-alanine, n-boc-n-methyl-l-alanine, s-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-l-alanine, boc-n-a-methyl-l-alanine, s-2-tert-butoxycarbonyl-methyl-amino-propionic acid, l-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino propanoic acid, 2s-2-tert-butoxy carbonyl methyl amino propanoic acidShow More | |
(2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid |
C9H17NO4 | |
VLHQXRIIQSTJCQ-LURJTMIESA-N | |
7009590 | |
CC(C(=O)O)N(C)C(=O)OC(C)(C)C |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only