missing translation for 'onlineSavingsMsg'
Learn More

BOC-N-Methyl-L-alanine, 99%, Thermo Scientific™

Chemical Identifiers

CAS 16948-16-6
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
InChI Key VLHQXRIIQSTJCQ-LURJTMIESA-N
Synonym boc-n-me-ala-oh, boc-n-methyl-l-alanine, n-boc-n-methyl-l-alanine, s-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-l-alanine, boc-n-a-methyl-l-alanine, s-2-tert-butoxycarbonyl-methyl-amino-propionic acid, l-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino propanoic acid, 2s-2-tert-butoxy carbonyl methyl amino propanoic acid
PubChem CID 7009590
IUPAC Name (2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
SMILES CC(C(=O)O)N(C)C(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442930010
SDS View Documents
Thermo Scientific Chemicals
442930010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC442930050
SDS View Documents
Thermo Scientific Chemicals
442930050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

16948-16-6
203.24
boc-n-me-ala-oh, boc-n-methyl-l-alanine, n-boc-n-methyl-l-alanine, s-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-l-alanine, boc-n-a-methyl-l-alanine, s-2-tert-butoxycarbonyl-methyl-amino-propionic acid, l-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino propanoic acid, 2s-2-tert-butoxy carbonyl methyl amino propanoic acid
(2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
C9H17NO4
VLHQXRIIQSTJCQ-LURJTMIESA-N
7009590
CC(C(=O)O)N(C)C(=O)OC(C)(C)C

Specifications

16948-16-6
Glass Bottle
1g
boc-n-me-ala-oh, boc-n-methyl-l-alanine, n-boc-n-methyl-l-alanine, s-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-l-alanine, boc-n-a-methyl-l-alanine, s-2-tert-butoxycarbonyl-methyl-amino-propionic acid, l-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino propanoic acid, 2s-2-tert-butoxy carbonyl methyl amino propanoic acid
CC(C(=O)O)N(C)C(=O)OC(C)(C)C
203.24
203.24
BOC-N-Methyl-L-alanine
99%
C9H17NO4
− 29.80
VLHQXRIIQSTJCQ-LURJTMIESA-N
(2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
7009590
99%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only