missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOC-3-iodo-L-alanine methyl ester, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 438460010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
BOC-3-iodo-L-alanine methyl ester | |
Authentic | |
Glass Bottle | |
MFCD00216579 | |
− 3.40 | |
UGZBFCCHLUWCQI-LURJTMIESA-N | |
methyl (2R)-3-iodo-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate | |
10903591 | |
97% |
93267-04-0 | |
96% min. (HPLC) | |
C9H16INO4 | |
1g | |
boc-beta-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, l-n-boc-3-iodoalanine methyl ester, l-n-boc-3-iodoalanine methyl ester, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome, a-iodo-ala-ome | |
COC(=O)[C@H](CI)NC(=O)OC(C)(C)C | |
329.13 | |
329.14 |
Chemical Identifiers
93267-04-0 | |
329.13 | |
UGZBFCCHLUWCQI-LURJTMIESA-N | |
10903591 | |
COC(=O)[C@H](CI)NC(=O)OC(C)(C)C |
C9H16INO4 | |
MFCD00216579 | |
boc-beta-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, l-n-boc-3-iodoalanine methyl ester, l-n-boc-3-iodoalanine methyl ester, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome, a-iodo-ala-omeShow More | |
methyl (2R)-3-iodo-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only