missing translation for 'onlineSavingsMsg'
Learn More

BOC-3-iodo-L-alanine methyl ester, 97%, Thermo Scientific™

$269.01 - $957.33

Chemical Identifiers

CAS 93267-04-0
Molecular Formula C9H16INO4
Molecular Weight (g/mol) 329.13
MDL Number MFCD00216579
InChI Key UGZBFCCHLUWCQI-LURJTMIESA-N
Synonym boc-beta-iodo-ala-ome, boc-3-iodo-l-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, n-tert-butoxycarbonyl-3-iodo-l-alanine methyl ester, l-n-boc-3-iodoalanine methyl ester, r-methyl 2-tert-butoxycarbonylamino-3-iodopropanoate, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate
PubChem CID 10903591
IUPAC Name methyl (2R)-3-iodo-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES COC(=O)[C@H](CI)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC438460010
View Documents Promotion Details
N/A 1g
Each for $269.01
Only null left
 
AC438460050
View Documents Promotion Details
N/A 5g
Each for $957.33
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

93267-04-0
329.13
UGZBFCCHLUWCQI-LURJTMIESA-N
10903591
COC(=O)[C@H](CI)NC(=O)OC(C)(C)C
C9H16INO4
MFCD00216579
boc-beta-iodo-ala-ome, boc-3-iodo-l-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, n-tert-butoxycarbonyl-3-iodo-l-alanine methyl ester, l-n-boc-3-iodoalanine methyl ester, r-methyl 2-tert-butoxycarbonylamino-3-iodopropanoate, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate
methyl (2R)-3-iodo-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate

Specifications

93267-04-0
96% min. (HPLC)
C9H16INO4
− 3.40
UGZBFCCHLUWCQI-LURJTMIESA-N
methyl (2R)-3-iodo-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
10903591
97%
Authentic
Glass Bottle
MFCD00216579
boc-beta-iodo-ala-ome, boc-3-iodo-l-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, n-tert-butoxycarbonyl-3-iodo-l-alanine methyl ester, l-n-boc-3-iodoalanine methyl ester, r-methyl 2-tert-butoxycarbonylamino-3-iodopropanoate, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate
COC(=O)[C@H](CI)NC(=O)OC(C)(C)C
329.13
329.14
BOC-3-iodo-L-alanine methyl ester
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only