missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%

Manufacturer:  Alfa Aesar™ H5556206

 View more versions of this product

Catalog No. AAH5556206



Description

Description

Applications
Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.

Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications

Specifications

Bis(2,2,2-trifluoroethyl) phosphite
76°C (169°F)
43°C to 44°C (2mmHg)
92466-70-1
(CF3CH2O)2P(O)H
bis 2,2,2-trifluoroethyl hydrogen phosphite, bis 2,2,2-trifluoroethyl phosphite, bis 2,2,2-trifluoroethyl phosphite, technical grade, bis 2,2,2-trifluoroethyl phosphonate, bis trifluoroethyl phosphonate, oxo-bis 2,2,2-trifluoroethoxy phosphanium, oxobis 2,2,2-trifluoroethoxy-??-phosphanylium, phosphonic acid bis 2,2,2-trifluoroethyl ester, phosphonic acid, bis 2,2,2-trifluoroethyl ester
C(C(F)(F)F)O[P+](=O)OCC(F)(F)F
245.037
246.05
90%
1.545 g/mL
1.332
5g
C4H4F6O3P+
MFCD00063314
IMDCVAFSSZPRRM-UHFFFAOYSA-N
oxo-bis(2,2,2-trifluoroethoxy)phosphanium
6386648
Liquid
Technical

Chemical Identifiers

92466-70-1
245.037
IMDCVAFSSZPRRM-UHFFFAOYSA-N
6386648
C(C(F)(F)F)O[P+](=O)OCC(F)(F)F
C4H4F6O3P+
MFCD00063314
bis 2,2,2-trifluoroethyl hydrogen phosphite, bis 2,2,2-trifluoroethyl phosphite, bis 2,2,2-trifluoroethyl phosphite, technical grade, bis 2,2,2-trifluoroethyl phosphonate, bis trifluoroethyl phosphonate, oxo-bis 2,2,2-trifluoroethoxy phosphanium, oxobis 2,2,2-trifluoroethoxy-??-phosphanylium, phosphonic acid bis 2,2,2-trifluoroethyl ester, phosphonic acid, bis 2,2,2-trifluoroethyl ester
oxo-bis(2,2,2-trifluoroethoxy)phosphanium
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents