missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%

Catalog No. p-7052736
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5556203 1 g
AAH5556206 5 g
2 options

Catalog No. AAH5556203

Supplier: Thermo Scientific Chemicals H5556203

Only null left

CAS: 92466-70-1 | C4H4F6O3P+ | 245.037 g/mol

Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.

Notes
Store at room temperature. Avoid strong oxidizing agents.

Chemical Identifiers

CAS 92466-70-1
Molecular Formula C4H4F6O3P+
Molecular Weight (g/mol) 245.037
MDL Number MFCD00063314
InChI Key IMDCVAFSSZPRRM-UHFFFAOYSA-N
Synonym bis 2,2,2-trifluoroethyl phosphonate, phosphonic acid bis 2,2,2-trifluoroethyl ester, oxo-bis 2,2,2-trifluoroethoxy phosphanium, phosphonic acid, bis 2,2,2-trifluoroethyl ester, bis trifluoroethyl phosphonate, bis 2,2,2-trifluoroethyl phosphite, bis 2,2,2-trifluoroethyl hydrogen phosphite, oxobis 2,2,2-trifluoroethoxy-??-phosphanylium, bis 2,2,2-trifluoroethyl phosphite, technical grade
PubChem CID 6386648
IUPAC Name oxo-bis(2,2,2-trifluoroethoxy)phosphanium
SMILES C(C(F)(F)F)O[P+](=O)OCC(F)(F)F

Specifications

Density 1.545 g/mL
Boiling Point 43°C to 44°C (2 mmHg)
Flash Point 76°C (169°F)
Refractive Index 1.332
Linear Formula (CF3CH2O)2P(O)H
Quantity 1 g
Formula Weight 246.05
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Bis(2,2,2-trifluoroethyl) phosphite
Show More Show Less
missing translation for 'healthHazard1' H227-H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
missing translation for 'healthHazard3' P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only