missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Lactose, Anhydrous, 99%, Spectrum™ Chemical

L3147, 5965-66-2, C12H22O11
$586.97 - $586.97
Chemical Identifiers
CAS | 5965-66-2 |
---|---|
Molecular Formula | C12H22O11 |
Molecular Weight (g/mol) | 342.30 |
MDL Number | MFCD00064521 |
InChI Key | GUBGYTABKSRVRQ-DCSYEGIMSA-N |
IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
SMILES | OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18600939
|
Spectrum Chemical Mfg Cor
L3147100GM |
100 g | Amber Glass Bottle |
Each for $586.97
|
|
|||||
Description
Spectrum™ Chemical beta-Lactose, Anhydrous , also known as milk sugar or 4-O-Β-D-galactopyranosyl-D-glucose, is a disaccharide sugar and is added to pills as a filler because of its physical properties. Anhydrous products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
5965-66-2 | |
342.30 | |
GUBGYTABKSRVRQ-DCSYEGIMSA-N | |
OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1OShow More |
C12H22O11 | |
MFCD00064521 | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triolShow More |
Specifications
5965-66-2 | |
Amber Glass Bottle | |
MFCD00064521 | |
GUBGYTABKSRVRQ-DCSYEGIMSA-N | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol | |
99% |
100% | |
C12H22O11 | |
100 g | |
OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O | |
342.30 | |
Reagent |