missing translation for 'onlineSavingsMsg'
Learn More

beta-Lactose, Anhydrous, 99%, Spectrum™ Chemical

Catalog No. 18600939
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600939 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18600939 Supplier Spectrum Chemical Mfg Cor Supplier No. L3147100GM
Only null left

L3147, 5965-66-2, C12H22O11

Spectrum™ Chemical beta-Lactose, Anhydrous , also known as milk sugar or 4-O-Β-D-galactopyranosyl-D-glucose, is a disaccharide sugar and is added to pills as a filler because of its physical properties. Anhydrous products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 5965-66-2
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00064521
InChI Key GUBGYTABKSRVRQ-DCSYEGIMSA-N
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 99%
Grade Reagent