Learn More
Avantorā„¢ BAKERBOND Speediskā„¢ Extraction Disks
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Extraction Disks are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
Supplier:  Avantorā„¢ 805706
 View more versions of this product
Catalog No. 02002378
Only null left
Explore available promotions
Description
Description
  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Diquat, Paraquat, EPA Method 549.1
C18 XF
20/Pk.
Expanded Extraction Station (8095-06)
Octyl (C8) Extraction Disk
50 mm
Product Suggestions
Product Suggestions